Learn More
iQ Biosciences Wistar Rat Splenocytes, 50M cells/vial
Catalog No. NC1345591
Encompass_Preferred
High-quality, high-viability (typically > 70%) splenocytes from the Wistar rat, a widely-used outbred rat strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.
Catalog No. NC1345591 Supplier iQ Biosciences Supplier No. IQBRSP202
High-quality, high-viability (typically > 70%) splenocytes from the Wistar rat, a widely-used outbred rat strain. Used for a wide variety of immunology-based applications, including in-vitro immunoassays and ex-vivo applications. All orders come with an iQ Certificate of Analysis.