Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Wisteria Floribunda (Japanese Wisteria) Lectin (WFL), fluorescein (FITC)

Catalog No. L32481
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32481 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32481 Supplier Invitrogen™ Supplier No. L32481
Only null left
Add to Cart
Add to Cart

Bright-green WFA-fluorophor conjugate

  • Wisteria Floribunda Lectin (WFA, WFL), fluorescein (FITC), is a bright-green WFA-fluorophor conjugate. WFA is sourced from Wisteria floribunda (common name: Japanese wisteria), which is a species of flowering plant in the pea family Fabaceae, native to Japan. Due to the specificity of WFA, it has been used in many fields to label certain cells.
  • In neurobiology, it has been used to detect perineuronal net. In the gastrointestinal tract, it stains the foveolar epithelium of normal gastric mucosa. This lectin has also been used as a marker of certain types of ovarian cancer.
  • Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: N-acetylgalactosamine
  • Inhibiting/eluting sugar: 200 mM N-acetylgalactosamine
  • Blood group specify: non-specific
  • Unconjugated lectin molecular weight: 116 KDa
  • Immunofluorescence working concentration: 5–20 μg/mL
  • WFA-FITC is supplied at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide. 0.1 mM Ca2+
  • A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mg
Source Wisteria floribunda
Conjugate Fluorescein
Content And Storage When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Label Type Classic Dyes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Subtype Lectins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.