Learn More
Bioworld WISTERIA FLORIBUNDA - LECTIN
Supplier:  Bioworld 215100121
Encompass
Wisteria floribunda Lectin isolated from Wisteria floribunda seeds. Specifications: Primary sugar specificity: N-acetylgalactosamine; Preferential binding activity: terminal N-acetylgalactosamine alpha or beta linked to the 1 o...
Catalog No. 50-198-746