Learn More
Bachem Ac-Trp-Glu-His-Asp-AMC
SDP
Supplier:  Bachem 4027662.0005
Encompass_Preferred
Ac-Trp-Glu-His-Asp-AMC, 189275-74-9, C38H40N8O11, Mr 784.78, 5mg. Ac-WEHD-AMC. With a kcat/Km of 3.3 · 106 M-1s-1 the fluorogenic caspase-1 substrate Ac-WEHD-AMC is cleaved 50-fold more efficiently thanAc-YVAD-AMC. This result indicates that the optimal tetrapeptide recognition sequence for caspase-1 is Trp-Glu-His-Asp and not Tyr-Val-Ala-Asp.
Catalog No. 50-194-6684