Learn More
Medchemexpress LLC HY-N2065 1mg Medchemexpress, Withaferin A CAS:5119-48-2 Purity:>98%
SDP
Catalog No. 501933880
Encompass_Preferred
Medchemexpress, HY-N2065 1mg Withaferin A CAS:5119-48-2 Withaferin A is a steroidal lactone isolated from Withania somnifera, inhibits NF-kB activation and targets vimentin , with potent antiinflammatory and anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3880 Supplier Medchemexpress LLC Supplier No. HYN20651MG
Medchemexpress, HY-N2065 1mg Withaferin A CAS:5119-48-2 Withaferin A is a steroidal lactone isolated from Withania somnifera, inhibits NF-kB activation and targets vimentin , with potent antiinflammatory and anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.