Learn More
Selleck Chemical LLC Withaferin A
SDP
Catalog No. 5000001761
Encompass_Preferred
Withaferin A (WFA) potently inhibits NF- B activation by preventing the tumor necrosis factor-induced activation of I B kinase via a thioalkylation-sensitive redox mechanism Withaferin A binds to the intermediate filament (IF) protein vimentin with antitumor and antiangiogenesis activity Withaferin A is a steroidal lactone isolated from Withania somnifera
Catalog No. 50-000-01761 Supplier Selleck Chemical LLC Supplier No. S8587100MG
Withaferin A (WFA) potently inhibits NF- B activation by preventing the tumor necrosis factor-induced activation of I B kinase via a thioalkylation-sensitive redox mechanism Withaferin A binds to the intermediate filament (IF) protein vimentin with antitumor and antiangiogenesis activity Withaferin A is a steroidal lactone isolated from Withania somnifera