Learn More
Selleck Chemical LLC Withaferin A
Supplier: Selleck Chemical LLC S85875MG
Withaferin A (WFA) potently inhibits NF- B activation by preventing the tumor necrosis factor-induced activation of I B kinase via a thioalkylation-sensitive redox mechanism Withaferin A binds to the intermediate filament (IF) protein vimentin with antitumor and antiangiogenesis activity Withaferin A is a steroidal lactone isolated from Withania somnifera