Learn More
Medchemexpress LLC HY-N2065 5mg , Withaferin A CAS:5119-48-2 Purity:>98%
SDP
Catalog No. 502260317
Encompass_Preferred
HY-N2065 5mg Withaferin A CAS: 5119-48-2 Withaferin A is a steroidal lactone isolated from Withania somnifera, inhibits NF-kB activation and targets vimentin, with potent antiinflammatory and anticancer activities. Withaferin A is an inhibitor of endothelial protein C receptor (EPCR) shedding. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0317 Supplier Medchemexpress LLC Supplier No. HYN20655MG
HY-N2065 5mg Withaferin A CAS: 5119-48-2 Withaferin A is a steroidal lactone isolated from Withania somnifera, inhibits NF-kB activation and targets vimentin, with potent antiinflammatory and anticancer activities. Withaferin A is an inhibitor of endothelial protein C receptor (EPCR) shedding. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.