Learn More
TARGETMOL CHEMICALS INC WITHAFERIN A 5MG
SDP
Catalog No. 502579740
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WITHAFERIN A is a novel class of NFkappaB inhibitors which hold promise as novel anti-inflammatory agents for treatment of various inflammatory disorders and/or cancer. purity: 99%
Catalog No. 50-257-9740 Supplier TARGETMOL CHEMICALS INC Supplier No. T56875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WITHAFERIN A is a novel class of NFkappaB inhibitors which hold promise as novel anti-inflammatory agents for treatment of various inflammatory disorders and/or cancer. purity: 99%