Learn More
Abcam Withaferin A from Withania somnifera, 1MG
Catalog No. 502707398
Encompass_Preferred
MW 470.6 g/mol, Purity >98%. Naturally occurring withanolide/steroidal lactone with anticancer properties. Demonstrates a number of cellular effects including the covalent binding to annexin II, inhibition of chymotrypsin-like activity, PKC and caspase 3. Causes translocation of cyctochrome C to the nucleous and the cleavage of PLC-γ1. Down-regulates Notch 1, 3, cdc25C, phosphorylated Akt and bcl-2 proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7398 Supplier Abcam Supplier No. AB1206441MG
Add to Cart
Add to Cart
MW 470.6 g/mol, Purity >98%. Naturally occurring withanolide/steroidal lactone with anticancer properties. Demonstrates a number of cellular effects including the covalent binding to annexin II, inhibition of chymotrypsin-like activity, PKC and caspase 3. Causes translocation of cyctochrome C to the nucleous and the cleavage of PLC-γ1. Down-regulates Notch 1, 3, cdc25C, phosphorylated Akt and bcl-2 proteins.
The product is subject to the following: Abcam Restricted Use Statement