Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Withanoside IV | 362472-81-9 | 1 MG
SDP

Catalog No. 5000417350
Encompass_Preferred

Withanoside IV is a steroid product isolated from the herbs of Withania somnifera. It is supplied as an off-white solid with a purity of 98.47% and is intended for research use only.

  • Molecular formula: C40H62O15
  • Molecular weight: 782.91
  • Purity: 98.47%
  • Appearance: Off-white solid
  • Storage conditions: -20°C, protect from light (solid); -80°C for 6 months or -20°C for 1 month (in solvent), protect from light
  • Soluble in DMSO (50 mg/mL)

Catalog No. 50-004-17350 Supplier Medchemexpress LLC Supplier No. HYN86931MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Withanoside IV is a steroid product isolated from the herbs of Withania somnifera. It is supplied as an off-white solid with a purity of 98.47% and is intended for research use only.

  • Molecular formula: C40H62O15
  • Molecular weight: 782.91
  • Purity: 98.47%
  • Appearance: Off-white solid
  • Storage conditions: -20°C, protect from light (solid); -80°C for 6 months or -20°C for 1 month (in solvent), protect from light
  • Soluble in DMSO (50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.