Learn More
Cayman Chemical WIthnolIde A 10mg
SDP
Catalog No. 502953705
Encompass_Preferred
A natural product that has antioxidant and neuroprotective activity; promotes neurite outgrowth in cultured neurons and in rodents injected with Aβ(25-35); recovered Aβ(25-35)-induced memory deficit in mice; reverses hypoxia-mediated neurodegeneration by restoring hypoxia induced glutathione depletion in the hippocampus
Catalog No. 50-295-3705 Supplier Cayman Chemical Supplier No. 1179710 MG
A natural product that has antioxidant and neuroprotective activity; promotes neurite outgrowth in cultured neurons and in rodents injected with Aβ(25-35); recovered Aβ(25-35)-induced memory deficit in mice; reverses hypoxia-mediated neurodegeneration by restoring hypoxia induced glutathione depletion in the hippocampus