Learn More
Mitegen Block, Wizard Cryo 1 and 2, 96 formulations, 1.7mL volumes
SDP
Catalog No. NC0462730
Encompass_Preferred
The Wizard Cryo line of random sparse matrix screens is designed for scientists who want to avoid the additional step of optimizing a cryoprotectant condition. Every Wizard Cryo formulation flash-freezes to a clear, amorphous glass in liquid nitrogen or in a cryo-stream at 100K. Crystals can be frozen directly from their growth drops, avoiding the additional step of pre-equilibration with an artificial cryo-condition that can damage the crystal. Eleven different cryocrystallants and sparing use of glycerol ensures a broad sampling of possible cryo conditions. Choose from Wizard Cryo 1 or 2 formulations in tubes or Wizard Cryo 1 and 2 together in a 96-well matrix block.
Catalog No. NC0462730 Supplier Mitegen Supplier No. 1008649
The Wizard Cryo line of random sparse matrix screens is designed for scientists who want to avoid the additional step of optimizing a cryoprotectant condition. Every Wizard Cryo formulation flash-freezes to a clear, amorphous glass in liquid nitrogen or in a cryo-stream at 100K. Crystals can be frozen directly from their growth drops, avoiding the additional step of pre-equilibration with an artificial cryo-condition that can damage the crystal. Eleven different cryocrystallants and sparing use of glycerol ensures a broad sampling of possible cryo conditions. Choose from Wizard Cryo 1 or 2 formulations in tubes or Wizard Cryo 1 and 2 together in a 96-well matrix block.