Learn More
Mitegen WIZARD CLASSIC 1&2 1.7ML VOL
SDP
Catalog No. NC0463232
Encompass_Preferred
Wizard Classic crystallization screen series. The Wizard™ Classic line of random sparse matrix screens is designed to increase your probability of producing crystals during the coarse screening phase when crystallizing biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).
Catalog No. NC0463232 Supplier Mitegen Supplier No. 1008647
Wizard Classic crystallization screen series. The Wizard™ Classic line of random sparse matrix screens is designed to increase your probability of producing crystals during the coarse screening phase when crystallizing biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).