Learn More
Mitegen Tubes, Wizard Cryo 1 and 2 Bundle, 96 formulations, 10mL volumes
SDP
Catalog No. NC0956217
Encompass_Preferred
The Wizard Cryo™ line of random sparse matrix screens is designed for scientists who want to avoid the additional step of optimizing a cryoprotectant condition. Every Wizard Cryo formulation flash-freezes to a clear, amorphous glass in liquid nitrogen or in a cryo-stream at 100K. Crystals can be frozen directly from their growth drops, avoiding the additional step of pre-equilibration with an artificial cryo-condition that can damage the crystal. Eleven different cryocrystallants and sparing use of glycerol ensures a broad sampling of possible cryo conditions. Choose from Wizard Cryo 1 or 2 formulations in tubes or Wizard Cryo 1 and 2 together in a 96-well matrix block.
Catalog No. NC0956217 Supplier Mitegen Supplier No. 1009538
The Wizard Cryo™ line of random sparse matrix screens is designed for scientists who want to avoid the additional step of optimizing a cryoprotectant condition. Every Wizard Cryo formulation flash-freezes to a clear, amorphous glass in liquid nitrogen or in a cryo-stream at 100K. Crystals can be frozen directly from their growth drops, avoiding the additional step of pre-equilibration with an artificial cryo-condition that can damage the crystal. Eleven different cryocrystallants and sparing use of glycerol ensures a broad sampling of possible cryo conditions. Choose from Wizard Cryo 1 or 2 formulations in tubes or Wizard Cryo 1 and 2 together in a 96-well matrix block.