Learn More
Mitegen Block, Wizard Cubic LCP, 96 formulations, 1.7mL volumes
SDP
Catalog No. NC0947541
Encompass_Preferred
The Wizard Cubic™ LCP Screen random sparse matrix is designed for the crystallization of biological macromolecules that are embedded within a lipidic cubic phase (LCP) host matrix. Every formulation in this 96-well plate is compatible with monoolein-based lipidic cubic phases.96 formulations, 1.7 mL volumes in a 96 well block plate.
Catalog No. NC0947541 Supplier Mitegen Supplier No. 1008650
The Wizard Cubic™ LCP Screen random sparse matrix is designed for the crystallization of biological macromolecules that are embedded within a lipidic cubic phase (LCP) host matrix. Every formulation in this 96-well plate is compatible with monoolein-based lipidic cubic phases.96 formulations, 1.7 mL volumes in a 96 well block plate.