Learn More
Mitegen Block, Wizard Precipitant Synergy Set, 192 formulations, 1.7mL volumes
SDP
Catalog No. NC0486534
Encompass_Preferred
Discover more protein crystallization hits with this unique screen by employing synergistic effects of mechanistically distinct precipitating agents (salts, organic solvents, polyethylene glycols, additives, pH). The Wizard Precipitant Synergy crystallization consists of 192 formulations, to be used for initial protein crystallization screening and crystallization optimization. The application of this screen is advised in particular for protein targets that have failed to yield hits with traditional random sparse matrix crystallization approaches.
Catalog No. NC0486534 Supplier Mitegen Supplier No. 1008652
Add to Cart
Add to Cart
Discover more protein crystallization hits with this unique screen by employing synergistic effects of mechanistically distinct precipitating agents (salts, organic solvents, polyethylene glycols, additives, pH). The Wizard Precipitant Synergy crystallization consists of 192 formulations, to be used for initial protein crystallization screening and crystallization optimization. The application of this screen is advised in particular for protein targets that have failed to yield hits with traditional random sparse matrix crystallization approaches.