Learn More
Bellco Glass, Inc. Water Jacketed Low Profile Over Head Drive Vessel W/4 SA,3L
SDP
Catalog No. 501944748
Encompass_Preferred
Ideal for use with water baths for precise temperature control. MADE IN THE USA Water jacketed low profile OHD vessel with:size: 3lfour (4) vertical 45mm sidearmsdimensions: 300mm od x 280mm overall heighttwo (2) #4 hose connectionsmaterial: type 1 class a borosilicate glasssupply: 100mm pp blue cap & white silicone liner & four 45mm pp blue caps & pouring rings
Catalog No. 50-194-4748 Supplier Bellco Glass, Inc. Supplier No. 196403440
Ideal for use with water baths for precise temperature control. MADE IN THE USA Water jacketed low profile OHD vessel with:size: 3lfour (4) vertical 45mm sidearmsdimensions: 300mm od x 280mm overall heighttwo (2) #4 hose connectionsmaterial: type 1 class a borosilicate glasssupply: 100mm pp blue cap & white silicone liner & four 45mm pp blue caps & pouring rings