Learn More
Bellco Glass, Inc. Water Jacketed Low Profile Over Head Drive Vessel W/6 Side Arms, 3L
SDP
Catalog No. 501944750
Encompass_Preferred
Ideal for use with water baths for precise temperature control. Supplied with 100mm center cap, sidearms cap, and pouring ring. 6 VERTICAL 45mm SIDEARMS. Material: type 1 class a Borosilicate glass. MADE IN THE USA Dimensions: 300mm od x 280mm overall height Two (2) #4 hose connection
Catalog No. 50-194-4750 Supplier Bellco Glass, Inc. Supplier No. 196403660
Ideal for use with water baths for precise temperature control. Supplied with 100mm center cap, sidearms cap, and pouring ring. 6 VERTICAL 45mm SIDEARMS. Material: type 1 class a Borosilicate glass. MADE IN THE USA Dimensions: 300mm od x 280mm overall height Two (2) #4 hose connection