Learn More
Bellco Glass, Inc. Water Jacketed Low Profile Over Head Drive Vessel W/6 Side Arms, 6 L
SDP
Catalog No. 501944763
Encompass_Preferred
Ideal for use with water baths for precise temperature control. Supplied with 100mm center cap, sidearms cap, and pouring ring. MADE IN THE USASpecifications:#4 Hose Barbs For 1/2″ Id TubingMaterial: Type 1 Class A Borosilicate GlassSupplied: 45mm Polypropylene Blue Cap & Pouring RingSize 6LOD x OAH: 300x325mm
Catalog No. 50-194-4763 Supplier Bellco Glass, Inc. Supplier No. 196406660
Ideal for use with water baths for precise temperature control. Supplied with 100mm center cap, sidearms cap, and pouring ring. MADE IN THE USASpecifications:#4 Hose Barbs For 1/2″ Id TubingMaterial: Type 1 Class A Borosilicate GlassSupplied: 45mm Polypropylene Blue Cap & Pouring RingSize 6LOD x OAH: 300x325mm