Learn More
Thermo Scientific™ WKITS™ Water QC Kit 1002
Catalog No. R4741002 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3/Pk.
Catalog No. Quantity
R4741002 3/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R4741002 Supplier Thermo Scientific™ Supplier No. R4741002
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ WKIT Water QC Kit 1002 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
Thermo Scientific™ WKIT Water QC Kit 1002 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
  • WaterBugs™ deliver <50 CFUs in a single inoculum to mimic water samples
Ordering Alerts:
Reagents are sufficient for one test/reconstituted vial.
Includes:
Contents: 9 preserved organism vials (3 per organism); 9 rehydration vials. ;Escherichia coli; strain C1, ;Enterobacter cloacae; ATCC™ 13047™,;Proteus mirabilis; ATCC™ 12453™.
TRUSTED_SUSTAINABILITY
Specifications
Description Water QC Kit 1002
Format WKITS
Product Type Quality Control Kit
Quantity 3/Pk.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.