Learn More
Cayman Chemical WKYMVm (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502947727
Encompass_Preferred
A synthetic peptide agonist of FPR1 and FPR2; induces calcium mobilization in ETFR cells transfected with FPR1 or FPR2 at 0.1 nM; stimulates chemotaxis of monocytes, dendritic cells, and NK cells, as well as superoxide production in monocytes and neutrophils, in vitro; reverses mucosal destruction, decreases in body weight, and colonic shortening in a mouse model of DSS-induced colitis at 8 mg/kg; topical application stimulates angiogenesis and accelerates re-epithelialization and granulation tissue formation in a rat model of diabetic cutaneous wounds
Catalog No. 50-294-7727 Supplier Cayman Chemical Supplier No. 3358910 MG
A synthetic peptide agonist of FPR1 and FPR2; induces calcium mobilization in ETFR cells transfected with FPR1 or FPR2 at 0.1 nM; stimulates chemotaxis of monocytes, dendritic cells, and NK cells, as well as superoxide production in monocytes and neutrophils, in vitro; reverses mucosal destruction, decreases in body weight, and colonic shortening in a mouse model of DSS-induced colitis at 8 mg/kg; topical application stimulates angiogenesis and accelerates re-epithelialization and granulation tissue formation in a rat model of diabetic cutaneous wounds