Learn More
Cole-Parmer Davis Instruments 6345 Leaf/Soil Moisture/Temp Monitoring Station; w/o Sensors
Catalog No. 50628432 Shop All Cole-Parmer Products
Encompass_Preferred
The Wireless Leaf & Soil Moisture/Temperature Station for Vantage Pro2™ monitors up to four temperature compensated Soil Moisture Sensors and two Leaf Wetness Sensors anywhere within the transmission range of your wireless Vantage Pro2 console. The station communicates directly to your Wireless Vantage Pro2 console/receiver over any one of eight user-selectable ID codes, and has a transmitting range of between 200' to 1000' (75 to 300 m) depending upon the environment. The station is solar powered, and includes a battery backup.
Catalog No. 50-628-432 Supplier Cole-Parmer Supplier No. 3204503
The Wireless Leaf & Soil Moisture/Temperature Station for Vantage Pro2™ monitors up to four temperature compensated Soil Moisture Sensors and two Leaf Wetness Sensors anywhere within the transmission range of your wireless Vantage Pro2 console. The station communicates directly to your Wireless Vantage Pro2 console/receiver over any one of eight user-selectable ID codes, and has a transmitting range of between 200' to 1000' (75 to 300 m) depending upon the environment. The station is solar powered, and includes a battery backup.