Learn More
Brew Plate™ WL NUTRIENT AGAR - 2KG

Description
Uses:
- Cultivating yeasts, molds, and bacteria
Specifications
Specifications
| Content And Storage | 2-30° C |
| Product Type | Agar |
| For Use With (Application) | Cultivating yeast and bacteria found in brewing. |
| Quantity | 1 EA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.