Learn More
NEOGEN WL Nutrient Agar, 500 g
SDP
Catalog No. 502015492
Encompass_Preferred
W-L Nutrient Agar is used for the cultivation of yeasts, molds, and bacteria encountered in brewing and industrial fermentations. W-L Nutrient Agar is not intended for use in the diagnosis of disease or other conditions in humans. W-L Nutrient Medium was developed by Green and Gray while studying various fermentation processes. An exhaustive study examining methods of fermentation control procedures in worts, beers, liquid yeasts and similar fermentation products led to the development of W-L Nutrient Medium. At a pH of 5.5, counts of viable baker’s yeast will grow on W-L Nutrient Medium. W-L Nutrient Medium is also referred to as “Wallerstein Laboratory Medium”.For more information visit https://www.neogen.com/solutions/Microbiology/wl-nutrient-agar
Catalog No. 50-201-5492 Supplier NEOGEN Supplier No. 700004506
W-L Nutrient Agar is used for the cultivation of yeasts, molds, and bacteria encountered in brewing and industrial fermentations. W-L Nutrient Agar is not intended for use in the diagnosis of disease or other conditions in humans. W-L Nutrient Medium was developed by Green and Gray while studying various fermentation processes. An exhaustive study examining methods of fermentation control procedures in worts, beers, liquid yeasts and similar fermentation products led to the development of W-L Nutrient Medium. At a pH of 5.5, counts of viable baker’s yeast will grow on W-L Nutrient Medium. W-L Nutrient Medium is also referred to as “Wallerstein Laboratory Medium”.For more information visit https://www.neogen.com/solutions/Microbiology/wl-nutrient-agar