Learn More
Selleck Chemical LLC WM-3835-5mg
SDP
Catalog No. 502389827
Encompass_Preferred
WM-3835 is a novel and high-specific small molecule Lysine Acetyltransferase 7 (KAT7, MYST2, HBO1) inhibitor, able to potently suppressed OS cell proliferation and migration, and leads to apoptosis activation.
Catalog No. 50-238-9827 Supplier Selleck Chemical LLC Supplier No. S98055MG
WM-3835 is a novel and high-specific small molecule Lysine Acetyltransferase 7 (KAT7, MYST2, HBO1) inhibitor, able to potently suppressed OS cell proliferation and migration, and leads to apoptosis activation.