Learn More
Medchemexpress LLC HY-134901 5mg , WM-3835 CAS:2229025-70-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1349015MG
Encompass_Preferred
HY-134901 5mg WM-3835 CAS: 2229025-70-9 WM-3835 is a potent and high-specific HBO1 (KAT7 or MYST2) inhibitor and binds directly to the acetyl-CoA binding site of HBO1 33. WM-3835 activates apoptosis while inhibits osteosarcoma (OS) cell proliferation, migration and invasion. WM-3835 has antitumor activity and potently inhibits pOS-1 xenograft growth in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0761