Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Wm-586 | 2894832-05-2 | C20H20F3N5O3S | 5 MG
SDP

Catalog No. 5000376242
Encompass_Preferred

WM-586 is a covalent WDR5 inhibitor that specifically decreases WDR5 and MYC interaction with an IC50 of 101 nM. It is used in research on various cancers, including neuroblastoma, breast cancer, bladder cancer, and colorectal cancer. It is for research use only.

  • Specifically decreases WDR5 and MYC interaction with an IC50 of 101 nM
  • Used in research on various cancers
  • Purity of 98.74%
  • Molecular weight: 467.46
  • Solid appearance
  • White to light yellow color

Catalog No. 50-003-76242 Supplier Medchemexpress LLC Supplier No. HY1537285MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

WM-586 is a covalent WDR5 inhibitor that specifically decreases WDR5 and MYC interaction with an IC50 of 101 nM. It is used in research on various cancers, including neuroblastoma, breast cancer, bladder cancer, and colorectal cancer. It is for research use only.

  • Specifically decreases WDR5 and MYC interaction with an IC50 of 101 nM
  • Used in research on various cancers
  • Purity of 98.74%
  • Molecular weight: 467.46
  • Solid appearance
  • White to light yellow color

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.