Learn More
Medchemexpress LLC WM-586 | 2894832-05-2 | C20H20F3N5O3S | 50 MG
SDP
Catalog No. 5000376873
Encompass_Preferred
WM-586 is a covalent WDR5 inhibitor that specifically decreases WDR5 and MYC interaction with an IC50 of 101 nM. It is used in research on various cancers, including neuroblastoma, breast cancer, bladder cancer, and colorectal cancer.
  • Specifically reduces interaction between WDR5 and MYC
  • Does not affect the binding of WDR5 to RBBP5
Catalog No. 50-003-76873 Supplier Medchemexpress LLC Supplier No. HY15372850MG
Add to Cart
Add to Cart
WM-586 is a covalent WDR5 inhibitor that specifically decreases WDR5 and MYC interaction with an IC50 of 101 nM. It is used in research on various cancers, including neuroblastoma, breast cancer, bladder cancer, and colorectal cancer.
  • Specifically reduces interaction between WDR5 and MYC
  • Does not affect the binding of WDR5 to RBBP5