Learn More
TARGETMOL CHEMICALS INC WM-8014 5MG
SDP
Catalog No. 502576772
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WM-8014 (MOZ-IN-3) is an inhibitor of MOZ(IC50=55 nM) a member of histone acetyltransferases. purity: 99%
Catalog No. 50-257-6772 Supplier TARGETMOL CHEMICALS INC Supplier No. T43625MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WM-8014 (MOZ-IN-3) is an inhibitor of MOZ(IC50=55 nM) a member of histone acetyltransferases. purity: 99%