Learn More
Andwin Scientific WM BOTTLE .5 GAL HDPE 2L 6CS
SDP
Catalog No. NC9344531
Encompass_Preferred
The Nalgene Large-Capacity Round Bottles are made of High-Density Polyethylene (HDPE) and are ideal for storing dry chemicals or water sampling. This can be utilized in freezer temperature down to -100C. It has a volume capacity of 2 liters or 1/2 gallon. This product has a wide mouth and has an included polypropylene screw closure. It is sold in a case of 6.
Catalog No. NC9344531 Supplier Andwin Scientific Supplier No. EF14767A
The Nalgene Large-Capacity Round Bottles are made of High-Density Polyethylene (HDPE) and are ideal for storing dry chemicals or water sampling. This can be utilized in freezer temperature down to -100C. It has a volume capacity of 2 liters or 1/2 gallon. This product has a wide mouth and has an included polypropylene screw closure. It is sold in a case of 6.