Learn More
Enterprise Technology Solutions WNA Comet Forks, Plant Starch, 7"L, 50/PK, Natural White
SDP
Catalog No. 5000789785
Encompass_Preferred
Forks are made of plant starch material, a renewable resource, and offer strength and heat tolerance. They are ideal for hot foods and liquids. Heat tolerance is up to 200 degrees Fahrenheit. Reinforced handles of these 7" long forks provide extra strength. Cutlery is not recommended for microwaves and is not compostable.
Catalog No. 50-007-89785 Supplier Enterprise Technology Solutions Supplier No. WNAEPS002
Forks are made of plant starch material, a renewable resource, and offer strength and heat tolerance. They are ideal for hot foods and liquids. Heat tolerance is up to 200 degrees Fahrenheit. Reinforced handles of these 7" long forks provide extra strength. Cutlery is not recommended for microwaves and is not compostable.