Learn More
Medchemexpress LLC HY-112094 10mg Medchemexpress, WNK-IN-11 CAS:2123489-30-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11209410MG
Encompass_Preferred
Medchemexpress, HY-112094 10mg WNK-IN-11 CAS:2123489-30-3 WNK-IN-11 is an allosteric With-No-Lysine (WNK) kinase inhibitor, with an IC50 of 4 nM for WNK1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9020