Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Wnk-in-11-d3 | 2123483-49-6 | 99.8% | 465.41 g/mol | C21H18D3Cl2N5OS | 5 MG
SDP

Catalog No. 5000263520
Encompass_Preferred

WNK-IN-11-d3 is a deuterated, orally active, selective, and potent inhibitor of With-No-Lysine (WNK) family kinases. It is used in preclinical research to probe WNK kinase signaling and to study regulation of cardiovascular homeostasis; the deuterated label supports tracer and pharmacokinetic experiments.

  • Deuterated analog suitable for pharmacokinetic and tracer studies.
  • Selective inhibition of WNK family kinases to modulate signaling pathways.
  • Reported purity ~99.8% for reproducible experimental results.
  • Molecular formula C21H18D3Cl2N5OS; molecular weight 465.41 g/mol.
  • Supplied as a 5 mg quantity for small-scale research use.
  • Intended for research use only; not for human consumption.

Catalog No. 50-002-63520 Supplier Medchemexpress LLC Supplier No. HY112094S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WNK-IN-11-d3 is a deuterated, orally active, selective, and potent inhibitor of With-No-Lysine (WNK) family kinases. It is used in preclinical research to probe WNK kinase signaling and to study regulation of cardiovascular homeostasis; the deuterated label supports tracer and pharmacokinetic experiments.

  • Deuterated analog suitable for pharmacokinetic and tracer studies.
  • Selective inhibition of WNK family kinases to modulate signaling pathways.
  • Reported purity ~99.8% for reproducible experimental results.
  • Molecular formula C21H18D3Cl2N5OS; molecular weight 465.41 g/mol.
  • Supplied as a 5 mg quantity for small-scale research use.
  • Intended for research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.