Learn More
Carnabio Usa Inc WNK2/10UG/CATALYTIC
Catalog No. 507541027
Encompass
WNK2, Catalytic domain, Wild type, Amino Acid 166-489, NP_006639.3, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1027 Supplier Carnabio Usa Inc Supplier No. 0518010
WNK2, Catalytic domain, Wild type, Amino Acid 166-489, NP_006639.3, Expressed in Insect (sf21); 10microg