Learn More
Aviva Systems Biology Corp WNK2 Antibody - N-terminal region (ARP65290_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65290_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WNK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human WNK2. Alternate names: KIAA1760, NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-464