Learn More
Aviva Systems Biology Corp WNK3 Antibody - N-terminal region (ARP48493_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48493_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WNK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WNK3. Alternate names: KIAA1566, PRKWNK3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59842