Learn More
ABclonal Technology WNK3 Rabbit mAb
SDP
Catalog No. 5000006326
Encompass_Preferred
This gene encodes a protein belonging to the with no lysine family of serine-threonine protein kinases These family members lack the catalytic lysine in subdomain II and instead have a conserved lysine in subdomain I This family member functions as a positive regulator of the transcellular Ca2 transport pathway and it plays a role in the increase of cell survival in a caspase-3-dependent pathway Alternative splicing results in multiple transcript variants
Catalog No. 50-000-06326 Supplier ABclonal Technology Supplier No. A23551100UL
This gene encodes a protein belonging to the with no lysine family of serine-threonine protein kinases These family members lack the catalytic lysine in subdomain II and instead have a conserved lysine in subdomain I This family member functions as a positive regulator of the transcellular Ca2 transport pathway and it plays a role in the increase of cell survival in a caspase-3-dependent pathway Alternative splicing results in multiple transcript variants