Learn More
Medchemexpress LLC HY-100626 10mg Medchemexpress, WNK463 CAS:2012607-27-9 Purity:>98%
SDP
Catalog No. 502028631
Encompass_Preferred
Medchemexpress, HY-100626 10mg WNK463 CAS:2012607-27-9 WNK463 is an orally bioavailable pan-With-No-Lysine (K) (WNK)-kinase inhibitor with IC50s of 5 nM, 1 nM, 6 nM, and 9 nM for WNK1, WNK2, WNK3, and WNK4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8631 Supplier Medchemexpress LLC Supplier No. HY10062610MG
Medchemexpress, HY-100626 10mg WNK463 CAS:2012607-27-9 WNK463 is an orally bioavailable pan-With-No-Lysine (K) (WNK)-kinase inhibitor with IC50s of 5 nM, 1 nM, 6 nM, and 9 nM for WNK1, WNK2, WNK3, and WNK4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.