Learn More
Medchemexpress LLC HY-100626 1mg , WNK463 CAS:2012607-27-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006261MG
Encompass_Preferred
HY-100626 1mg WNK463 CAS: 2012607-27-9 WNK463 is an orally bioavailable pan-With-No-Lysine (K) (WNK)-kinase inhibitor with IC50s of 5 nM, 1 nM, 6 nM, and 9 nM for WNK1, WNK2, WNK3, and WNK4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9417