Learn More
Enzo Life Sciences Wnt-3a (mouse), (recombinant) (1 µg)
SDP
Supplier:  Enzo Life Sciences ENZ60001C001
Encompass_Preferred
Modulator of embryogenesis, cell proliferation, stem cell differentation, and bone, glucose and lipid metabolism. MW: ~40kDa (SDS-PAGE). Endotoxin content: Less than 0.2 ng per µg (2EU/µg). Purity: >75% by SDS-PAGE and >75% by HPLC analyses. Formulation: Lyophilized solid. Source: Cell culture. Use/Stability: Stable for at least 12 months after receipt when stored, as supplied, at -80°C. Following reconstitution, stable for three months when stored at or below -20°C. UniProt: P27467. Long Term Storage: -80°C
Catalog No. NC0852339