Learn More
Selleck Chemical LLC Wnt agonist 1 10mg 2095432-75-8 BML-284 HCL, BML-284 hydrochloride, AMBMP hydrochloride, AMBMP HCL
SDP
Catalog No. 501365750
Encompass_Preferred
Wnt agonist 1 (BML-284 HCL, AMBMP HCL) is a cell permeable Wnt signaling pathway activator, which induces ?-catenin- and TCF-dependent transcriptional activity with EC50 of 0.7 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5750 Supplier Selleck Chemical LLC Supplier No. S817810MG
Wnt agonist 1 (BML-284 HCL, AMBMP HCL) is a cell permeable Wnt signaling pathway activator, which induces ?-catenin- and TCF-dependent transcriptional activity with EC50 of 0.7 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.