Learn More
Apexbio Technology LLC Wnt agonist 1(Synonyms: Wnt Agonist I, Wnt signaling activator I, Wnt agonist compound 1, Wnt agonist-I), 200mg, CAS: 853220-52-7.
SDP
Catalog No. 5000060221
Encompass_Preferred
Wnt agonist 1 (CAS 853220-52-7) is a small-molecule stimulator of the canonical Wnt signaling pathway It functions by activating -catenin-dependent transcription mediated through the TCF transcription factor with an EC50 of approximately 0 7 M In Xenopus embryos treated from developmental stage 10 5 to stage 40 with Wnt agonist 1 (10 M) notable cephalic defects including reduced head size or absent eyes were observed mirroring phenotypes associated with increased Wnt signaling Researchers use Wnt agonist 1 to investigate Wnt pathway-regulated cellular differentiation and development
Catalog No. 50-000-60221 Supplier Apexbio Technology LLC Supplier No. B6059200
Wnt agonist 1 (CAS 853220-52-7) is a small-molecule stimulator of the canonical Wnt signaling pathway It functions by activating -catenin-dependent transcription mediated through the TCF transcription factor with an EC50 of approximately 0 7 M In Xenopus embryos treated from developmental stage 10 5 to stage 40 with Wnt agonist 1 (10 M) notable cephalic defects including reduced head size or absent eyes were observed mirroring phenotypes associated with increased Wnt signaling Researchers use Wnt agonist 1 to investigate Wnt pathway-regulated cellular differentiation and development