Learn More
Cayman Chemical Wnt Agonist I, Purity: ≥98%, CAS: 853220-52-7, 10 mg
SDP
Catalog No. 502985161
Encompass_Preferred
A cell-permeable activator of Wnt signaling that does not inhibit GSK3β (IC50 > 60 µM); induces nuclear accumulation of β-catenin, increasing transcriptional activity driven by TCF (EC50 = 700 nM)
Catalog No. 50-298-5161 Supplier Cayman Chemical Supplier No. 1990310 MG
A cell-permeable activator of Wnt signaling that does not inhibit GSK3β (IC50 > 60 µM); induces nuclear accumulation of β-catenin, increasing transcriptional activity driven by TCF (EC50 = 700 nM)