Learn More
Medchemexpress LLC Wnt/B-catenin agonis 10mM | 10MM 1ML
SDP
Catalog No. 5000229660
Encompass_Preferred
Wnt/ -catenin agonist 1 (compound 3f) is a Wnt/ -catenin signalling pathway agonist with an EC50 of 0 27 M[1]
Catalog No. 50-002-29660 Supplier Medchemexpress LLC Supplier No. HY11432110MM 1ML
Wnt/ -catenin agonist 1 (compound 3f) is a Wnt/ -catenin signalling pathway agonist with an EC50 of 0 27 M[1]