Learn More
Cell Signaling Technology Wnt/β-Catenin Activated Targets Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502730595
Encompass
The Wnt/β-Catenin Activated Targets Antibody Sampler Kit provides an economical solution for investigating target proteins within the Wnt/β-Catenin signaling pathway, which is implicated in vertebrate development and tumorigenesis. This kit includes a collection of primary antibodies to analyze downstream effectors including CD44, cyclin D1, c-Jun, LEF1, Met, MMP-7, c-Myc, and TCF1/TCF7. Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide. Store at -20°C. Do not aliquot.
  • Contains enough primary antibody to perform at least 2 western blots per target.
  • Includes HRP-linked anti-mouse and anti-rabbit IgG secondary antibodies for detection.
  • Validated for multiple applications including western blotting and immunoprecipitation.
  • Includes 8 specific primary antibodies for canonical pathway targets.
Catalog No. 50-273-0595 Supplier Cell Signaling Technology Supplier No. 8655T
The Wnt/β-Catenin Activated Targets Antibody Sampler Kit provides an economical solution for investigating target proteins within the Wnt/β-Catenin signaling pathway, which is implicated in vertebrate development and tumorigenesis. This kit includes a collection of primary antibodies to analyze downstream effectors including CD44, cyclin D1, c-Jun, LEF1, Met, MMP-7, c-Myc, and TCF1/TCF7. Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide. Store at -20°C. Do not aliquot.
  • Contains enough primary antibody to perform at least 2 western blots per target.
  • Includes HRP-linked anti-mouse and anti-rabbit IgG secondary antibodies for detection.
  • Validated for multiple applications including western blotting and immunoprecipitation.
  • Includes 8 specific primary antibodies for canonical pathway targets.