Learn More
Medchemexpress LLC HY-15659 10mg , Wnt-C59 CAS:1243243-89-1 Purity:>98%
SDP
Catalog No. 502260934
Encompass_Preferred
HY-15659 10mg Wnt-C59 CAS: 1243243-89-1 Wnt-C59 (C59) is a highly potent and oral porcupine (PORCN) inhibitor with an IC50 of 74 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0934 Supplier Medchemexpress LLC Supplier No. HY1565910MG
HY-15659 10mg Wnt-C59 CAS: 1243243-89-1 Wnt-C59 (C59) is a highly potent and oral porcupine (PORCN) inhibitor with an IC50 of 74 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.