Learn More
Medchemexpress LLC HY-15659 100mg , Wnt-C59 C59 CAS:1243243-89-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15659100MG
Encompass_Preferred
Medchemexpress, HY-15659 100mg Wnt-C59 C59 CAS:1243243-89-1 Formula:C25H21N3O PORCN Wnt Purity:98% Wnt-C59 is a potent PORCN enzymatic activity and Wnt inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3076