Learn More
Apexbio Technology LLC Wnt-C59 1243243-89-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062411
Encompass_Preferred
Wnt-C59 (CAS 1243243-89-1) is a selective inhibitor of the Wnt signaling pathway targeting Porcupine (PORCN) an acyltransferase essential for Wnt protein secretion Exhibiting an IC50 of 74 pM Wnt-C59 blocks cellular secretion of Wnt ligands thus preventing downstream activation of Wnt-dependent transcriptional activity Studies using cell lines such as HT1080 HeLa and cholangiocarcinoma-derived CC-LP-1 SUN-1079 and CC-SW-1 have demonstrated diminished cell viability reduced proliferation and increased apoptosis upon Wnt-C59 treatment In WNT-driven tumor mouse models Wnt-C59 administration significantly inhibits tumor growth highlighting its utility for studying Wnt-mediated cancer pathogenesis and therapeutic intervention strategies
Catalog No. 50-000-62411 Supplier Apexbio Technology LLC Supplier No. A86855.1
Wnt-C59 (CAS 1243243-89-1) is a selective inhibitor of the Wnt signaling pathway targeting Porcupine (PORCN) an acyltransferase essential for Wnt protein secretion Exhibiting an IC50 of 74 pM Wnt-C59 blocks cellular secretion of Wnt ligands thus preventing downstream activation of Wnt-dependent transcriptional activity Studies using cell lines such as HT1080 HeLa and cholangiocarcinoma-derived CC-LP-1 SUN-1079 and CC-SW-1 have demonstrated diminished cell viability reduced proliferation and increased apoptosis upon Wnt-C59 treatment In WNT-driven tumor mouse models Wnt-C59 administration significantly inhibits tumor growth highlighting its utility for studying Wnt-mediated cancer pathogenesis and therapeutic intervention strategies