Learn More
Cellagen Technology WNT-C59,2MG
Supplier:  Cellagen Technology C76412S
Encompass
Wnt-C59 is a potent selective Wnt signaling inhibitor. It has IC50 less than 0.11 nM in Wnt-Luc reporter assay, preventing palmitylation of Wnt proteins by Porcupine, thereby blocking Wnt secretion and activity.
Catalog No. NC0186427